Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001106809.4 | XP_001106809.1 | 219 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |