Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMPPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265981.1 | NP_001252910.1 | 699 | 103 | ||
XM_015130792.2 | XP_014986278.1 | 807 | 103 | |||
GenBank mRNA | JV046884.1 | - | 699 | 103 | ||
JV681944.1 | - | 778 | 103 |
Gene Symbol: RNF123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU346973.1 | - | 4464 | 103 | ||
JU476736.1 | - | 5005 | 103 | |||
JU476737.1 | - | 4924 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |