Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14H11orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115477.2 | XP_014970963.1 | 3112 | 104 | ||
XM_015115478.2 | XP_014970964.1 | 392 | 104 | |||
XM_015115479.2 | XP_014970965.1 | 401 | 104 | |||
GenBank mRNA | JV649238.1 | - | 104 | |||
JV712999.1 | - | 302 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |