Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266715.1 | NP_001253644.1 | 534 | 93 | ||
XM_015131757.2 | XP_014987243.1 | 1261 | 93 | |||
GenBank mRNA | JU322506.1 | - | 526 | 93 | ||
JU473727.1 | - | 534 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |