Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015144514.2 | XP_015000000.2 | 517 | 151 | ||
XM_028851851.1 | XP_028707684.1 | 423 | 151 | |||
XM_028851852.1 | XP_028707685.1 | 423 | 151 | |||
XM_028851853.1 | XP_028707686.1 | 423 | 151 | |||
XM_028851854.1 | XP_028707687.1 | 452 | 151 | |||
XM_028851855.1 | XP_028707688.1 | 517 | 151 | |||
XM_028851860.1 | XP_028707693.1 | 423 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |