Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRRS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194376.2 | NP_001181305.2 | 477 | 79 | ||
GenBank mRNA | JU327908.1 | - | 140 | 79 | ||
JV611618.1 | - | 321 | 79 | |||
JV678852.1 | - | 392 | 79 |