Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032937.1 | NP_001028109.1 | 650 | 106 | ||
GenBank mRNA | AY457046.1 | - | 650 | 106 | ||
JU322273.1 | - | 928 | 106 | |||
JU470879.1 | - | 856 | 106 | |||
JV567653.1 | - | 938 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |