Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257751.2 | NP_001244680.1 | 195 | 154 | ||
GenBank mRNA | JU334042.1 | - | 195 | 154 | ||
JU473308.1 | - | 242 | 154 | |||
JV046784.1 | - | 242 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |