Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001089477.4 | XP_001089477.1 | 911 | 137 | ||
GenBank mRNA | JU337254.1 | - | 858 | 137 | ||
JU477960.1 | - | 855 | 137 | |||
JV599223.1 | - | 894 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |