Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123697.2 | XP_014979183.2 | 4304 | 133 | ||
XM_028839234.1 | XP_028695067.1 | 4040 | 133 | |||
XM_028839235.1 | XP_028695068.1 | 4040 | 133 | |||
XM_028839236.1 | XP_028695069.1 | 4184 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |