Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JADE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100171.4 | XP_001100171.3 | 352 | 101 | ||
XM_002806209.3 | XP_002806255.2 | 592 | 101 | |||
XM_028842510.1 | XP_028698343.1 | 489 | 101 | |||
XM_028842511.1 | XP_028698344.1 | 463 | 101 | |||
GenBank mRNA | JU337313.1 | - | 364 | 101 | ||
JU478015.1 | - | 294 | 101 | |||
JV048826.1 | - | 447 | 101 | |||
JV666670.1 | - | 450 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |