Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIEZO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028841099.1 | XP_028696932.1 | 2257 | 88 | ||
XM_028841100.1 | XP_028696933.1 | 2257 | 88 |