Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266236.1 | NP_001253165.1 | 868 | 158 | ||
XM_015136457.2 | XP_014991943.1 | 1130 | 158 | |||
XM_028846741.1 | XP_028702574.1 | 1101 | 158 | |||
GenBank mRNA | JU322749.1 | - | 1046 | 158 | ||
JU471796.1 | - | 868 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |