Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEPROTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266983.1 | NP_001253912.1 | 1070 | 151 | ||
GenBank mRNA | JU320962.1 | - | 124 | 151 | ||
JU320963.1 | - | 124 | 151 | |||
JU474945.1 | - | 252 | 151 | |||
JU474946.1 | - | 252 | 151 | |||
JV046597.1 | - | 1070 | 151 | |||
JV576690.1 | - | 150 | 151 | |||
JV700478.1 | - | 613 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |