Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015109755.2 | XP_014965241.2 | 382 | 117 | ||
XM_015109757.2 | XP_014965243.2 | 473 | 117 | |||
XM_028830252.1 | XP_028686085.1 | 351 | 117 | |||
GenBank mRNA | JU322824.1 | - | 349 | 117 | ||
JV070530.1 | - | 347 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |