Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015142296.2 | XP_014997782.2 | 695 | 87 | ||
XM_015142297.2 | XP_014997783.2 | 199 | 87 | |||
GenBank mRNA | JU330633.1 | - | 52 | 87 | ||
JV711721.1 | - | 73 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |