Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193804.2 | NP_001180733.1 | 994 | 154 | ||
XM_015145508.2 | XP_015000994.1 | 709 | 154 | |||
XM_015145509.2 | XP_015000995.1 | 741 | 154 | |||
XM_028852127.1 | XP_028707960.1 | 748 | 154 | |||
XM_028852128.1 | XP_028707961.1 | 748 | 154 | |||
GenBank mRNA | JV043440.1 | - | 665 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |