Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141455.2 | XP_014996941.1 | 583 | 118 | ||
GenBank mRNA | JU322152.1 | - | 475 | 118 | ||
JV600700.1 | - | 475 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |