Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015111612.2 | XP_014967098.2 | 1749 | 83 | ||
XM_015111613.2 | XP_014967099.2 | 1710 | 83 | |||
XM_015111616.2 | XP_014967102.2 | 1749 | 83 | |||
XM_028832011.1 | XP_028687844.1 | 1815 | 83 | |||
GenBank mRNA | JV546840.1 | - | 1177 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |