Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SARAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145102.2 | XP_015000588.1 | 729 | 71 | ||
GenBank mRNA | JU322457.1 | - | 249 | 71 | ||
JU475967.1 | - | 251 | 71 | |||
JV045475.1 | - | 142 | 71 | |||
JV638187.1 | - | 247 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |