Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001111331.4 | XP_001111331.2 | 359 | 126 | ||
XM_015129973.2 | XP_014985459.2 | 461 | 228 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |