Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107634.4 | XP_001107634.1 | 814 | 84 | ||
GenBank mRNA | JV062980.1 | - | 132 | 84 | ||
JV653364.1 | - | 231 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |