Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIGD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015119890.2 | XP_014975376.1 | 519 | 124 | ||
XM_015119891.2 | XP_014975377.1 | 477 | 124 | |||
XM_028836606.1 | XP_028692439.1 | 368 | 124 | |||
XM_028836607.1 | XP_028692440.1 | 577 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |