Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMMDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194452.2 | NP_001181381.1 | 696 | 159 | ||
GenBank mRNA | JU323495.1 | - | 707 | 159 | ||
JU487800.1 | - | 735 | 159 | |||
JV045055.1 | - | 687 | 159 | |||
JV528414.1 | - | 720 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |