Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266669.1 | NP_001253598.1 | 176 | 97 | ||
XM_015146053.2 | XP_015001539.1 | 196 | 97 | |||
GenBank mRNA | JU322681.1 | - | 206 | 97 | ||
JU322682.1 | - | 206 | 97 | |||
JU470749.1 | - | 325 | 97 | |||
JU470750.1 | - | 325 | 97 | |||
JV045484.1 | - | 176 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |