Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP3A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040414.1 | NP_001035504.1 | 213 | 146 | ||
GenBank mRNA | AY334551.1 | - | 213 | 146 | ||
AY582531.1 | - | 213 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta Inventoried |