Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPN1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194626.3 | NP_001181555.3 | 325 | 98 | ||
GenBank mRNA | CD767084.1 | - | 262 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |