Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260523.1 | NP_001247452.1 | 980 | 81 | ||
XM_028832806.1 | XP_028688639.1 | 1087 | 81 | |||
XM_028832807.1 | XP_028688640.1 | 1087 | 81 | |||
GenBank mRNA | JU476111.1 | - | 974 | 81 | ||
JV044348.1 | - | 980 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |