Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265700.2 | NP_001252629.1 | 1595 | 148 | ||
XM_015110501.2 | XP_014965987.2 | 148 | ||||
XM_015110502.2 | XP_014965988.2 | 148 | ||||
XM_015110503.2 | XP_014965989.1 | 148 | ||||
GenBank mRNA | JU335130.1 | - | 1598 | 148 | ||
JU476130.1 | - | 1598 | 148 | |||
JV565141.1 | - | 1597 | 148 | |||
JV717606.1 | - | 1598 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |