Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265700.2 | NP_001252629.1 | 1851 | 119 | ||
XM_015110501.2 | XP_014965987.2 | 1979 | 119 | |||
XM_015110502.2 | XP_014965988.2 | 1936 | 119 | |||
XM_015110503.2 | XP_014965989.1 | 1979 | 119 | |||
GenBank mRNA | JU335130.1 | - | 1854 | 119 | ||
JU476130.1 | - | 1854 | 119 | |||
JV565141.1 | - | 1853 | 119 | |||
JV717606.1 | - | 1854 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |