Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015117794.2 | XP_014973280.1 | 650 | 127 | ||
XM_028834280.1 | XP_028690113.1 | 385 | 127 | |||
GenBank mRNA | JU323108.1 | - | 631 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |