Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM255A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085757.2 | XP_001085757.1 | 604 | 149 | ||
GenBank mRNA | JU335206.1 | - | 496 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |