Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEZF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028844307.1 | XP_028700140.1 | 1695 | 61 | ||
XM_028844308.1 | XP_028700141.1 | 1405 | 61 | |||
XM_028844309.1 | XP_028700142.1 | 1666 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |