Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265856.1 | NP_001252785.1 | 720 | 118 | ||
GenBank mRNA | JU335247.1 | - | 719 | 118 | ||
JU476218.1 | - | 686 | 118 | |||
JV047512.1 | - | 720 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |