Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DZANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149557.2 | XP_015005043.2 | 985 | 119 | ||
XM_015149559.2 | XP_015005045.2 | 1055 | 119 | |||
XM_028828148.1 | XP_028683981.1 | 1120 | 119 | |||
XM_028828149.1 | XP_028683982.1 | 1011 | 119 | |||
XM_028828150.1 | XP_028683983.1 | 1520 | 119 | |||
XM_028828151.1 | XP_028683984.1 | 1502 | 119 | |||
XM_028828152.1 | XP_028683985.1 | 1577 | 119 | |||
XM_028828153.1 | XP_028683986.1 | 1577 | 119 | |||
XM_028828154.1 | XP_028683987.1 | 1577 | 119 | |||
XM_028828156.1 | XP_028683989.1 | 1577 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |