Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257752.1 | NP_001244681.1 | 1378 | 158 | ||
GenBank mRNA | JU335273.1 | - | 1378 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |