Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139049.2 | XP_014994535.2 | 360 | 109 | ||
XM_015139050.2 | XP_014994536.2 | 302 | 109 | |||
XM_015139051.2 | XP_014994537.2 | 360 | 109 | |||
XM_015139053.2 | XP_014994539.2 | 360 | 109 | |||
XM_015139054.2 | XP_014994540.2 | 360 | 109 | |||
XM_015139055.2 | XP_014994541.2 | 360 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |