Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257809.1 | NP_001244738.1 | 1041 | 138 | ||
XM_015130537.2 | XP_014986023.1 | 1003 | 138 | |||
XM_028843080.1 | XP_028698913.1 | 1257 | 138 | |||
GenBank mRNA | JU335324.1 | - | 1041 | 138 | ||
JU335325.1 | - | 1041 | 138 | |||
JU335326.1 | - | 1041 | 138 | |||
JU335327.1 | - | 1041 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |