Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261587.2 | NP_001248516.1 | 138 | 76 | ||
GenBank mRNA | JU322989.1 | - | 143 | 76 | ||
JV044129.1 | - | 138 | 76 | |||
JV668702.1 | - | 192 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |