Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265707.1 | NP_001252636.1 | 291 | 89 | ||
XM_015127975.2 | XP_014983461.1 | 268 | 89 | |||
XM_015127976.1 | XP_014983462.1 | 232 | 89 | |||
XM_028841777.1 | XP_028697610.1 | 207 | 89 | |||
GenBank mRNA | JU325184.1 | - | 163 | 89 | ||
JV636269.1 | - | 291 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |