Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASNSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194770.3 | NP_001181699.2 | 1773 | 116 | ||
XM_001106875.3 | XP_001106875.2 | 1847 | 116 | |||
XM_001106941.4 | XP_001106941.2 | 1880 | 116 | |||
XM_028830549.1 | XP_028686382.1 | 1742 | 116 | |||
GenBank mRNA | JU323128.1 | - | 1843 | 116 | ||
JU472280.1 | - | 1853 | 116 | |||
JV047437.1 | - | 1744 | 116 | |||
JV703919.1 | - | 1891 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |