Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261255.1 | NP_001248184.1 | 746 | 137 | ||
XM_015152396.2 | XP_015007882.1 | 779 | 137 | |||
XM_015152397.2 | XP_015007883.1 | 463 | 137 | |||
XM_015152398.2 | XP_015007884.1 | 477 | 137 | |||
GenBank mRNA | JU322193.1 | - | 732 | 137 | ||
JU471425.1 | - | 746 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |