Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001115919.4 | XP_001115919.1 | 1077 | 96 | ||
XM_028829656.1 | XP_028685489.1 | 1084 | 96 | |||
GenBank mRNA | JU325648.1 | - | 148 | 96 | ||
JV656376.1 | - | 147 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |