Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115228.2 | XP_014970714.1 | 2707 | 104 | ||
XM_015115229.2 | XP_014970715.1 | 2704 | 104 | |||
XM_015115230.2 | XP_014970716.1 | 2680 | 104 | |||
XM_015115232.2 | XP_014970718.1 | 2677 | 104 | |||
XM_015115233.2 | XP_014970719.1 | 2554 | 104 | |||
XM_015115234.2 | XP_014970720.1 | 2542 | 104 | |||
XM_015115236.2 | XP_014970722.1 | 2527 | 104 | |||
XM_028833834.1 | XP_028689667.1 | 2707 | 104 | |||
GenBank mRNA | JU335540.1 | - | 2658 | 104 | ||
JU476467.1 | - | 2658 | 104 | |||
JV719156.1 | - | 2663 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |