Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015111197.2 | XP_014966683.1 | 1105 | 68 | ||
XM_015111199.2 | XP_014966685.1 | 1105 | 68 | |||
XM_015111201.2 | XP_014966687.1 | 932 | 68 | |||
XM_015111202.2 | XP_014966688.1 | 757 | 68 | |||
XM_028831165.1 | XP_028686998.1 | 932 | 68 | |||
XM_028831166.1 | XP_028686999.1 | 757 | 68 | |||
GenBank mRNA | JV066164.1 | - | 562 | 68 | ||
JV656427.1 | - | 560 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |