Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266050.1 | NP_001252979.1 | 605 | 113 | ||
XM_028827283.1 | XP_028683116.1 | 1046 | 113 | |||
XM_028827284.1 | XP_028683117.1 | 837 | 113 | |||
XM_028827285.1 | XP_028683118.1 | 767 | 113 | |||
XM_028827286.1 | XP_028683119.1 | 1134 | 113 | |||
GenBank mRNA | JU321400.1 | - | 605 | 113 | ||
JV513791.1 | - | 507 | 113 | |||
JV656575.1 | - | 623 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |