Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257565.1 | NP_001244494.1 | 1085 | 61 | ||
GenBank mRNA | JU335621.1 | - | 1085 | 61 | ||
JU476535.1 | - | 1104 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |