Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257912.1 | NP_001244841.1 | 1063 | 110 | ||
XM_028843109.1 | XP_028698942.1 | 1749 | 110 | |||
GenBank mRNA | JU321422.1 | - | 1063 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |