Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028846505.1 | XP_028702338.1 | 4273 | 153 | ||
XM_028846507.1 | XP_028702340.1 | 4496 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |