Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265670.1 | NP_001252599.1 | 447 | 114 | ||
XM_028845750.1 | XP_028701583.1 | 1943 | 114 | |||
GenBank mRNA | JU325338.1 | - | 305 | 114 | ||
JV636317.1 | - | 447 | 114 | |||
JV721065.1 | - | 373 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |